- Synthesis and biological activities of novel structural analogues of 2-arachidonoylglycerol, an endogenous cannabinoid receptor ligand.
Synthesis and biological activities of novel structural analogues of 2-arachidonoylglycerol, an endogenous cannabinoid receptor ligand.
Bioorganic & medicinal chemistry letters (2001-07-17)
Y Suhara, S Nakane, S Arai, H Takayama, K Waku, Y Ishima, T Sugiura
PMID11454463
ZUSAMMENFASSUNG
Novel analogues of 2-arachidonoylglycerol (2-AG), an endogenous cannabinoid receptor ligand, were developed. Chemical synthesis of these analogues (2-AGA105 and 2-AGA109) was accomplished starting from 2-octyn-1-ol and diethyl malonate and employing Wittig coupling of triene phosphonate with an aldehyde intermediate in a convergent and stereoselective manner. These analogues should be useful lead compounds for the development of novel 2-AG mimetics.
MATERIALIEN