Merck
  • The effect of a bradykinin B2 receptor antagonist, NPC-567, on allergen-induced airway responses in a porcine model.
The effect of a bradykinin B2 receptor antagonist, NPC-567, on allergen-induced airway responses in a porcine model.
Inflammation research : official journal of the European Histamine Research Society ... [et al.] (2001-10-18)
H Sylvin, I van der Ploeg, K Alving
ZUSAMMENFASSUNG
In order to assess the effect of selective blocking of the bradykinin (BK) B2 receptor in allergic airway reactions, the BK B2 receptor antagonist NPC-567 was administered to sensitized pigs before allergen challenge. Fourteen specific pathogen-free pigs sensitized to Ascaris suum were used. NPC-567 (2.5 mg, in 1 ml saline) was delivered as an aerosol twice to six pigs. Ascaris antigen (in 2 ml saline) was given as an aerosol to all pigs and airway mechanics were monitored for 8 h. NPC-567 (2.5 mg) was given at t = -30 min (in 1ml saline) and mixed with the antigen at t = 0 to six pigs. Allergen challenge caused an acute reaction with a rapid, significant increase in airways resistance from 4.1 +/- 0.5 cm H2O/l/s to a maximum of 16.2 +/- 3.0 cm H2O/l/s in the control pigs. In the NPC-567-treated pigs, the resistance only increased from 2.9 +/- 0.3 cm H2O/l/s to 6.5 +/- 0.9 cm H2O/l/s (p<0.005 compared to controls). There was also a higher reduction in dynamic lung compliance in the controls than in the treated animals upon allergen challenge. The histamine concentration in urine in the control pigs was markedly elevated after allergen challenge peaking at 15-30 min. This release was inhibited in the NPC-567-treated pigs. The BK B2 receptor antagonist NPC-567 seems to be effective in inhibiting the acute response to allergen in the pig airways, possibly due to inhibition of mast cell activation via indirect mechanisms. The late obstructive response was reduced as well, probably as a consequence of the reduced mediator release in the acute reaction.