Merck
  • Synthesis and cytotoxic evaluation of alpha-methylene-gamma-butyrolactone bearing naphthalene and naphtho[2,1-b]furan derivatives.
Synthesis and cytotoxic evaluation of alpha-methylene-gamma-butyrolactone bearing naphthalene and naphtho[2,1-b]furan derivatives.
European journal of medicinal chemistry (2002-04-19)
Kuan-Han Lee, Bor-Ruey Huang
ZUSAMMENFASSUNG
Naphthalene alpha-methylene-gamma-butyrolactones exhibits a unique cytotoxicity profile. They are highly cytostatic for leukaemia cancer cells but are not cytocidal. For almost all the solid tumours tested, they are both cytostatic and cytocidal. Substitution of a bromo atom on either naphthalene or both naphthalene and gamma-phenyl moiety of the lactone enhanced potency while retaining the same cytotoxicity profile. The tricyclic naphtho[2,1-b]furan derivatives, prepared from 2-hydroxy-1-naphthaldehyde in an efficient pathway, also possess the same cytotoxicity profile.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
α-Methylen-γ-butyrolacton, 97%