- Excision of transposable elements from the chalcone isomerase and dihydroflavonol 4-reductase genes may contribute to the variegation of the yellow-flowered carnation (Dianthus caryophyllus).
Excision of transposable elements from the chalcone isomerase and dihydroflavonol 4-reductase genes may contribute to the variegation of the yellow-flowered carnation (Dianthus caryophyllus).
Plant & cell physiology (2002-06-01)
Yoshio Itoh, Daisuke Higeta, Akane Suzuki, Hiroyuki Yoshida, Yoshihiro Ozeki
PMID12040106
ZUSAMMENFASSUNG
In the "Rhapsody" cultivar of the carnation, which bears white flowers variegated with red flecks and sectors, a transposable element, dTdic1, belonging to the Ac/Ds superfamily, was found within the dihydroflavonol 4-reductase (DFR) gene. The red flecks and sectors of "Rhapsody" may be attributable to a reversion to DFR activity after the excision of dTdic1. The yellow color of the carnation petals is attributed to the synthesis and accumulation of chalcone 2'-glucoside. In several of the carnation cultivars that bear yellow flowers variegated with white flecks and sectors, both the chalcone isomerase (CHI) and DFR genes are disrupted by dTdic1.