Merck
  • [The biological fate of 4-butyl-4-(beta-carboxypropionyloxymethyl)-1,2-diphenyl-3,5-pyrazolidinedione(suxibuzone), antiinflammatory drug. I. Plasma levels and urinary excretion after oral administration of suxibuzone (author's transl)].
[The biological fate of 4-butyl-4-(beta-carboxypropionyloxymethyl)-1,2-diphenyl-3,5-pyrazolidinedione(suxibuzone), antiinflammatory drug. I. Plasma levels and urinary excretion after oral administration of suxibuzone (author's transl)].
Yakugaku zasshi : Journal of the Pharmaceutical Society of Japan (1979-12-01)
T Shindo, Y Yasuda, K Taira, N Mitani, A Kanda, A Akazawa
PMID120893