- [Critical study of the therapeutic effects of phlebotropic drugs on chronic venous insufficiency].
[Critical study of the therapeutic effects of phlebotropic drugs on chronic venous insufficiency].
Phlebologie (1979-10-01)
K Roztocil, I Prerovsky, I Oliva
PMID120945
ZUSAMMENFASSUNG
The authors describe two types of test. Their aim was to verify the therapeutic potency of drugs used in the treatment of chronic venous disorders. The tests were carried out using both a single prescription and using successive prescriptions. The results showed that with strict clinical surveillance, phlebotropic drugs had definite therapeutic effects both on the subjective disturbances and on certain objective parameters.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Tribenosid, European Pharmacopoeia (EP) Reference Standard