Merck
  • Importance of the methyl group at C10 of squalene for hopene biosynthesis and novel carbocyclic skeletons with 6/5 + 5/5 + (6) ring system(s).
Importance of the methyl group at C10 of squalene for hopene biosynthesis and novel carbocyclic skeletons with 6/5 + 5/5 + (6) ring system(s).
Organic letters (2002-07-19)
Tsutomu Hoshino, Shumi Ohashi
ZUSAMMENFASSUNG
[reaction: see text] Incubation of (6E,10E,14E,18E)-2,6,10,19,23-pentamethyl-tetracosa-2,6,10,14,18,22-hexaene with Alicyclobacillus acidocaldarius hopene cyclase afforded four products having two types of carbocyclic skeletons, i.e., two hopane products and two products having an unprecedented carbocyclic skeleton of 6/5 + 5/5 +6 pentacyclic and 6/5 + 5/5 tetracyclic ring systems. The former two hopane skeletons were formed from the bioconversion of C15-desmethylsqualene and the latter two skeletons from that of C10-desmethylsqualene.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Hop-22(29)-en -Lösung, 0.1 mg/mL in isooctane, analytical standard