Merck
  • Retinal pigment epithelial cells phagocytosis of T lymphocytes: possible implication in the immune privilege of the eye.
Retinal pigment epithelial cells phagocytosis of T lymphocytes: possible implication in the immune privilege of the eye.
The British journal of ophthalmology (2002-11-26)
F Willermain, L Caspers-Velu, B Nowak, P Stordeur, R Mosselmans, I Salmon, T Velu, C Bruyns
ZUSAMMENFASSUNG
To investigate the capability of retinal pigment epithelium (RPE) cells to phagocytose T lymphocytes and to further analyse the immunobiological consequences of this phagocytosis. Human RPE cells pretreated or not by cytochalasin, a phagocytosis inhibitor, were co-cultured with T lymphocytes for different time points. Phagocytosis was investigated by optic microscopy, electron microscopy, and flow cytometry. T cell proliferation was measured by (3)H thymidine incorporation. RPE interleukin 1beta mRNA expression was quantified by real time PCR. RPE cells phagocytose apoptotic and non-apoptotic T lymphocytes, in a time dependent manner. This is an active process mediated through actin polymerisation, blocked by cytochalasin E treatment. Inhibition of RPE cell phagocytosis capabilities within RPE-T cell co-cultures led to an increase of lectin induced T cell proliferation and an upregulation of interleukin 1beta mRNA expression in RPE cells. It is postulated that T lymphocyte phagocytosis by RPE cells might, by decreasing the total number of T lymphocytes, removing apoptotic lymphocytes, and downregulating the expression of IL-1beta, participate in vivo in the induction and maintenance of the immune privilege of the eye, preventing the development of intraocular inflammation.