Merck
  • [Kinetic parameters of hydrolysis of CpA and UpA sequences in an oligoribonucleotide by compounds functionally mimicking ribonuclease A].
[Kinetic parameters of hydrolysis of CpA and UpA sequences in an oligoribonucleotide by compounds functionally mimicking ribonuclease A].
Molekuliarnaia biologiia (2002-12-26)
N G Beloglazova, N L Mironova, D A Konevets, V A Petiuk, V N Sil'nikov, V V Vlasov, M A Zenkova
ZUSAMMENFASSUNG
Kinetic parameters of cleavage of CpA and UpA sequences in an oligoribonucleotide under the action of artificial ribonuclease ABL3C1 were measured. The compounds were built of RNA-binding domain B, catalytic fragment C, linker L3 comprising 3 methylene groups, and aliphatic fragment A. The rate of cleavage of phosphodiester bonds in CpA sequence within decaribonucleotide UUCAUGUAAA was shown to be 3.4 +/- 0.2 times higher than in UpA sequence. The rate of cleavage of phosphodiester bonds were found to depend on substrate length: a thousandfold increase in cleavage rate constant was observed for CpA sequence in decaribonucleotide as compared with diribonucleotide monophosphate CpA. A slight decrease in the cleavage rates was observed for the reactions proceeding in different buffers at pH 7.0: imidazole > HEPES > phosphate > cacodylate. At the same time, the ratio of cleavage rates for CpA and UpA sequences remained constant.