Merck
Hyperphenylalaninemia reduces creatine kinase activity in the cerebral cortex of rats.
International journal of developmental neuroscience : the official journal of the International Society for Developmental Neuroscience (2003-03-05)
Elisa Costabeber, Adriana Kessler, Carlos Severo Dutra-Filho, Angela Terezinha de Souza Wyse, Moacir Wajner, Clóvis Milton Duval Wannmacher
ZUSAMMENFASSUNG
Phenylketonuria (PKU) is a metabolic disorder accumulating phenylalanine (Phe) and its metabolites in plasma and tissues of the patients. Considering that phenylalanine is the main neurotoxic metabolite, and brain energy homeostasis seems to be affected in phenylketonuria, our main objective was to investigate the effect of acute and chronic hyperphenylalaninemia (HPA) on creatine kinase (CK) activity in brain cortex of Wistar rats. Hyperphenylalaninemia was induced by subcutaneous administration of 5.2 micromol phenylalanine + 2.4 micromol alpha-methylphenylalanine (phenylalanine hydroxylase (PAH) inhibitor)/g of body weight. We also investigated the in vitro effect of phenylalanine and/or alpha-methylphenylalanine on creatine kinase activity in the brain cortex of non-treated rats. The results showed that phenylalanine significantly inhibited creatine kinase activity in vitro and reduced the enzyme activity in vivo. Considering the importance of creatine kinase for the maintenance of energy homeostasis in brain, if this enzyme inhibition also occurs in phenylketonuric patients, it is possible that creatine kinase inhibition may be one of the mechanisms by which phenylalanine is neurotoxic in phenylketonuria.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
α-Methyl-DL-phenylalanin, 98%