Merck
  • Alternative and sources of reagents and supplies of fluorophore-assisted carbohydrate electrophoresis(FACE).
Alternative and sources of reagents and supplies of fluorophore-assisted carbohydrate electrophoresis(FACE).
Glycobiology (2003-03-28)
Mark A Lehrman, Ningguo Gao
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2-Aminoacridon, BioReagent, suitable for fluorescence, ≥98.0% (HPLC)