Merck
9-deaza-5'-noraristeromycin.
Bioorganic & medicinal chemistry (2003-07-03)
Meral Tuncbilek, Stewart W Schneller
ZUSAMMENFASSUNG
9-deaza-5'-noraristeromycin (2) has been prepared in 10 steps from the readily available (+)-(1R,4S)-4-t-butyldimethylsilyloxycyclopent-2-en-1-yl acetate. Compound 2 was evaluated against a large number of viruses. No activity was found nor did 2 display cyctotoxicity towards the viral host cells.