Merck
Novel synthesis of polymorphous nanocrystalline KNbO3 by a low temperature solution method.
Journal of nanoscience and nanotechnology (2003-08-12)
Jun-Feng Liu, Xiao-Lin Li, Ya-Dong Li
ZUSAMMENFASSUNG
Orthorhombic and rhombohedral single nanocrystalline KNbO3 with different morphologies was synthesized respectively at a relatively low temperature by simply changing the reaction solvent; a conceivable reaction process was illustrated based on the finding of a hexahedral intermediate product during the formation of KNbO3.