Merck
  • The in vitro substrate regiospecificity of recombinant UGT85B1, the cyanohydrin glucosyltransferase from Sorghum bicolor.
The in vitro substrate regiospecificity of recombinant UGT85B1, the cyanohydrin glucosyltransferase from Sorghum bicolor.
Phytochemistry (2003-08-30)
Karina Sinding Hansen, Charlotte Kristensen, David Bruce Tattersall, Patrik Raymond Jones, Carl Erik Olsen, Søren Bak, Birger Lindberg Møller
ZUSAMMENFASSUNG
The in vitro substrate specificity of UDP-glucose:p-hydroxymandelonitrile-O-glucosyltransferase from Sorghum bicolor (UGT85B1) was examined using a range of potential acceptor molecules, including cyanohydrins, terpenoids, phenolics, hexanol derivatives and plant hormones. Qualitative enzyme activity assays employing 20 different putative substrates were performed and 15 proved to be glucosylated using recombinant UGT85B1 isolated from Escherichia coli. K(m) and k(cat) values were determined for nine of these substrates including mandelonitrile, geraniol, nerol and beta-citronellol, 2-hydroxy-3-methoxybenzyl alcohol, 1-hexanol, cis-3-hexen-1-ol, 3-methyl-3-buten-1-ol and 3-methyl-2-buten-1-ol. UGT85B1 has a rather broad substrate specificity in vitro but shows regiospecificity, demanding the presence of a sterically unhindered hydroxyl group e.g. as part of a cyanohydrin function, as a primary alcohol or as a phenolic hydroxyl group and being influenced by the stereochemistry and/or interactive chemistry of the substituents on the hydroxyl-bearing carbon atom.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dhurrin, ≥95% (HPLC)