- A prevention trial on cerebral infarction with dl-alpha-tocopheryl nicotinate--rationales for trial. The Alpha-Tocopheryl Nicotinate Prevention Study Group.
A prevention trial on cerebral infarction with dl-alpha-tocopheryl nicotinate--rationales for trial. The Alpha-Tocopheryl Nicotinate Prevention Study Group.
Journal of nutritional science and vitaminology (1992-01-01)
Y Hata, A Terashi, T Matsuzaki, E Ohtomo, Y Goto
PMID1297741
MATERIALIEN