Merck
  • [ON SHORTENING THE TREATMENT TIME IN DYSENTERY PATIENTS WITH ANTIBIOTICS AND CHEMICAL PREPARATIONS].
[ON SHORTENING THE TREATMENT TIME IN DYSENTERY PATIENTS WITH ANTIBIOTICS AND CHEMICAL PREPARATIONS].
Antibiotiki (1963-09-01)
E G POPKOVA, N V IURKOV