Merck
Metabotropic glutamate receptor regulation of neuronal cell death.
Experimental neurology (2003-11-05)
Alison Berent Spillson, James W Russell
ZUSAMMENFASSUNG
The metabotropic glutamate receptors (mGluRs) are a family of glutamate-sensitive receptors that regulate neuronal function separately from the ionotropic glutamate receptors. By coupling to guanosine nucleotide-binding proteins (G proteins), mGluRs are able to regulate neuronal injury and survival, likely through a series of downstream protein kinase and cysteine protease signaling pathways that affect mitochondrial regulated programmed cell death (PCD). The physiological relevance of this system is supported by evidence that mGluRs are associated with cell survival in several central nervous system neurodegenerative diseases. Evidence is presented that mGluRs are also able to prevent PCD in the peripheral nervous system, and that this may provide a novel mechanism for treatment of diabetic neuropathy. In dorsal root ganglion (DRG) neurons, a high glucose load increases generation of reactive oxygen species (ROS), destabilizes the inner mitochondrial membrane potential (Deltapsi(M)), induces cytochrome c release from the mitochondrial intermembrane space, and induces downstream activation of caspases. In high-glucose conditions, the group II metabotropic glutamate agonist N-acetylaspartylglutamate (NAAG) blocks caspase activation and is completely reversed by the mGluR3 antagonist (S)-alpha-ethylglutamic acid (EGLU). Furthermore, the direct mGluR3 agonist (2R,4R)-4-aminopyrrolidine-2, 4-dicarboxylate (APDC) prevents induction of ROS. Together these findings are consistent with an emerging concept that mGluRs may protect against cellular injury by regulating oxidative stress in the neuron. More complete understanding of the complex PCD regulatory pathways mediated by mGluRs will provide new therapeutic approaches for the treatment of a wide variety of neurodegenerative diseases.