Merck
  • Investigation of the Origanum onites L. essential oil using the chorioallantoic membrane (CAM) assay.
Investigation of the Origanum onites L. essential oil using the chorioallantoic membrane (CAM) assay.
Journal of agricultural and food chemistry (2004-01-22)
Fatih Demirci, Dietrich H Paper, Gerhard Franz, K Hüsnü Can Başer
ZUSAMMENFASSUNG
The in vivo test on the chorioallantoic membrane of the fertilized hen's egg (CAM assay) is a current method to determine antiangiogenic, antiinflammatory activity and toxic effects of individual compounds or complex plant extracts. The method is used for testing natural compounds in small amounts for revealing various modes of action and the complex mechanisms related to angiogenesis and inflammation. Furthermore, possible side effects such as membrane irritation, toxic, and anticoagulant properties of the investigated material in question can be detected. For the evaluation, the essential oil obtained by hydrodistillation of the aerial parts of Origanum onites L., a common spice and medicinal plant, was tested for its effect in the chorioallantoic membrane (CAM) assay. The essential oil composition was revealed by means of gas chromatography-mass spectrometry (GC-MS). Eighty three components were identified, representing 99.1% of the total oil. Carvacrol, thymol, p-cymene, and gamma-terpinene were found as major components and were also individually tested in the CAM assay. Along with the monoterpenes carvacrol and thymol, their methyl ether derivatives were also examined for comparison of their physiological action. Neither the essential oil nor its components showed any pronounced antiinflammatory or antiangiogenic property in the CAM assay, at 10-250 microg/pellet. However, the irritant effect of the essential oil was linked to thymol in a dose-response fashion, up to 10 microg/pellet, where it was still showing irritation.