- A thin-layer chromatographic method to determine process impurities in leucovorin calcium.
A thin-layer chromatographic method to determine process impurities in leucovorin calcium.
Journal of pharmaceutical sciences (1992-12-01)
D White, P Varlashkin, D N Rusch
PMID1491341
ZUSAMMENFASSUNG
A densitometric thin-layer chromatographic method for the analysis of process impurities in leucovorin calcium was developed and validated. Using this method, folic acid, N10-formyldihydrofolic acid, p-aminobenzoic acid, and N-(4-aminobenzoyl)-L-glutamic acid were monitored with a limit of detection of approximately 0.1% each. Both absorbance and fluorescence densitometric evaluations were utilized; fluorescence evaluation selectively detected impurities that were not chromatographically resolved from leucovorin.