Merck
Profound insights into squalene cyclization.
Chemistry & biology (2004-04-29)
Karl Poralla
ZUSAMMENFASSUNG
In this issue of Chemistry & Biology, our understanding of the formation of pentacyclic hopene from the linear squalene is enhanced by an X-ray structure of a complex between squalene-hopene cyclase and the substrate analog 2-azasqualene.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Squalen, ≥98%, liquid
Supelco
Squalen, analytical standard
Supelco
Hop-22(29)-en -Lösung, 0.1 mg/mL in isooctane, analytical standard