Merck
  • Change in the circadian rhythm of blood pressure in pseudo-hypoaldosteronism type 2 in response to thiazide diuretics.
Change in the circadian rhythm of blood pressure in pseudo-hypoaldosteronism type 2 in response to thiazide diuretics.
Clinical nephrology (2004-06-09)
T Uzu, K Takahara, A Yamauchi