Merck
  • High-level expression of a neoagarobiose-producing beta-agarase gene from Agarivorans sp. JAMB-A11 in Bacillus subtilis and enzymic properties of the recombinant enzyme.
High-level expression of a neoagarobiose-producing beta-agarase gene from Agarivorans sp. JAMB-A11 in Bacillus subtilis and enzymic properties of the recombinant enzyme.
Biotechnology and applied biochemistry (2004-08-17)
Yukari Ohta, Yuji Hatada, Susumu Ito, Koki Horikoshi
ZUSAMMENFASSUNG
The structural gene for a neoagarobiose-producing beta-agarase of an Agarivorans isolate was expressed in Bacillus subtilis. High-level production of the recombinant enzyme was achieved corresponding to a level of 1.9 x 10(4) units/l of the culture broth. The efficiency of the production is thus 30-fold greater than that of the original strain. The recombinant enzyme (RagaA11) had a molecular mass of 105 kDa and a specific activity of 371 units/mg. The optimal pH and temperature of the enzyme were 7.5-8.0 and 40 degrees C respectively. The enzyme is an endo-type beta-agarase hydrolysing not only agarose, but also neoagarotetraose, to yield neoagarobiose as the final main product, representing approx. 90 mol% of total products. RagaA11 would be useful for industrial production of neoagarobiose.