- 3-Deazaadenosine, a stabilizer of whole-blood homocysteine content, does not interfere with the single-enzyme homocysteine assay while totally inhibiting the enzyme conversion homocysteine immunoassay.
3-Deazaadenosine, a stabilizer of whole-blood homocysteine content, does not interfere with the single-enzyme homocysteine assay while totally inhibiting the enzyme conversion homocysteine immunoassay.
Clinical chemistry (2004-08-28)
Qinghong Han, Xinghua Sun, Mingxu Xu, Nan Zhang, Li Tang, Yuying Tan, Robert M Hoffman
PMID15331515
MATERIALIEN