- Self-condensation of N-tert-butanesulfinyl aldimines: application to the rapid asymmetric synthesis of biologically important amine-containing compounds.
Self-condensation of N-tert-butanesulfinyl aldimines: application to the rapid asymmetric synthesis of biologically important amine-containing compounds.
Organic letters (2004-09-25)
Laurie B Schenkel, Jonathan A Ellman
PMID15387563
ZUSAMMENFASSUNG
[reaction: see text] Highly diastereoselective intra- and intermolecular self-condensation reactions of N-tert-butanesulfinyl aldimines have been developed and applied to the rapid, asymmetric synthesis of trans-2-aminocyclopentanecarboxylic acid and the drug candidate SC-53116. Key to both syntheses is a novel microwave-assisted reaction in which N-sulfinyl aldimines are cleanly converted into nitriles in high-yielding concerted elimination processes.