Merck
  • A short route to enantiomerically pure benzophenanthridinone skeleton: synthesis of lactone analogues of narciclasine and lycoricidine.
A short route to enantiomerically pure benzophenanthridinone skeleton: synthesis of lactone analogues of narciclasine and lycoricidine.
The Journal of organic chemistry (2004-09-25)
Said Ibn-Ahmed, Mustapha Khaldi, Françoise Chrétien, Yves Chapleur
ZUSAMMENFASSUNG
Condensation of functionalized o-toluamide anions on a carbohydrate-derived lactone, followed by intramolecular aldol cyclization, provides enantiomerically pure 2-arylcyclohexenones. Different approaches for the stereoselective transformation of the carbonyl group of these key intermediates into an amino group were unsuccessful. However 1,4-addition of thiolate and concomitant ring closure to isocoumarine provided a useful method for the transformation of the tertiary amide function. Opening of the isocoumarin with ammonia provided the corresponding amide and recovery of the enone system. Subsequent reductive amination of this cyclohexenone was found to depend on the nature of the protecting groups and led to the protected form of 4-epi- and -iso-narciclasine. Oxo analogues of narciclasine and epi-narciclasine and lycoricidine were also obtained after reduction of the enone and subsequent lactonization. They showed no biological activity as antitumor agents.