Merck
  • A highly flexible dinuclear ruthenium(II)-platinum(II) complex: crystal structure and binding to 9-ethylguanine.
A highly flexible dinuclear ruthenium(II)-platinum(II) complex: crystal structure and binding to 9-ethylguanine.
Angewandte Chemie (International ed. in English) (2004-10-21)
Karlijn van der Schilden, Fedora Garcìa, Huub Kooijman, Anthony L Spek, Jaap G Haasnoot, Jan Reedijk
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
9-Ethylguanine