Merck
Lead phosphate minerals: solubility and dissolution by model and natural ligands.
Environmental science & technology (2004-12-04)
Carmen Enid Martínez, Astrid R Jacobson, Murray B Mcbride
ZUSAMMENFASSUNG
Due to their relatively low solubility, lead-phosphate minerals may control Pb solution levels at a low value in natural environments. We reportthe solubility of Pb from two lead-orthophosphate mineral suspensions (beta-Pb9(PO4)6 and PbHPO4) after aging for 3 years. Lead (Pb2+) activity in the aged suspensions was compared to the activity calculated using the Ksp values of various Pb-PO4 minerals reported in the literature. We also determine the time-dependent dissolution of the aged lead-phosphate minerals by organic and inorganic ligands containing S-functional groups (cysteine, methionine, and thiosulfate) and by a soil extracted humic acid. We find the activity of Pb2+ in the aged lead-phosphate suspensions to be 1-2 orders of magnitude higher than predicted by the Ksp values reported in the literature. Disagreement between measured and Ksp-calculated activities has been reported in other investigations of Pb-PO4 minerals; we compiled some of the data and present them together with our results. Furthermore, the time-dependent dissolution experiments indicate that, in most cases, lead phosphates are partly dissolved in the presence of soluble ligands, i.e., model sulfides and humic acid. The soil-extracted humic acid enhanced the dissolution of Pb from the high pH (7.2) lead-phosphate (beta-Pb9(PO4)6) mineral while suppressing Pb dissolution from the low pH (3.8) lead-phosphate (PbHPO4) mineral. While the low molecularweight sulfur-containing ligands enhanced Pb dissolution, their effect was less pronounced. We conclude that (i) nonequilibrium conditions prevail in the mineral suspensions even after 3 years of aging; and (ii) soluble ligands present in soils, sediments, and natural waters can potentially dissolve Pb from lead-phosphate minerals; such ligands, then, may enhance the biological availability and mobility of Pb in the environment.