Merck
  • Synthesis and pharmacological evaluation of glycine-modified analogues of the neuroprotective agent glycyl-L-prolyl-L-glutamic acid (GPE).
Synthesis and pharmacological evaluation of glycine-modified analogues of the neuroprotective agent glycyl-L-prolyl-L-glutamic acid (GPE).
Bioorganic & medicinal chemistry (2004-12-16)
Michelle Y H Lai, Margaret A Brimble, David J Callis, Paul W R Harris, Mark S Levi, Frank Sieg
ZUSAMMENFASSUNG
The synthesis of 10 G*PE analogues, wherein the glycine residue has been modified, is described by coupling readily accessible dibenzyl-L-prolyl-L-glutamate 2 with various analogues of glycine. Pharmacological evaluation of the novel compounds was undertaken to further understand the role of the glycine residue on the observed neuroprotective properties of the endogenous tripeptide GPE.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Gly-Pro-Glu, ≥98% (HPLC)