Merck
  • Synthesis and biological evaluation of new phenidone analogues as potential dual cyclooxygenase (COX-1 and COX-2) and human lipoxygenase (5-LOX) inhibitors.
Synthesis and biological evaluation of new phenidone analogues as potential dual cyclooxygenase (COX-1 and COX-2) and human lipoxygenase (5-LOX) inhibitors.
Farmaco (Societa chimica italiana : 1989) (2005-01-18)
Claudia Cusan, Giampiero Spalluto, Maurizio Prato, Michael Adams, Antje Bodensieck, Rudolf Bauer, Aurelia Tubaro, Paolo Bernardi, Tatiana Da Ros
ZUSAMMENFASSUNG
A new series of potential human 5-LOX inhibitors structurally related to the 1-phenyl-3-pyrazolidinone (phenidone, 2) has been synthesized and the activity against COX-1, COX-2, and human 5-LOX enzymes has been evaluated. In contrast with literature data, we observed that phenidone resulted to be inactive against human 5-LOX, while retains its activity against cyclooxygenases in a micromolar range. The present results suggest that the substitution of the amino function at the 4-position is detrimental in terms of activity toward COX-1 and COX-2, while the presence of a double bond at the 4,5-position does not alter the biological profile against COX. The absence of activity vs. human 5-LOX strongly suggests a re-consideration of phenidone and its analogs as 5-LOX inhibitors in humans.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
1-Phenyl-3-pyrazolidinon, 97%