Merck
Adulteration of South African traditional herbal remedies.
Therapeutic drug monitoring (2005-01-25)
Tracy Snyman, Michael J Stewart, Alida Grove, Vanessa Steenkamp
ZUSAMMENFASSUNG
Traditional herbal remedies are used as alternative medicine by a large proportion of people worldwide. Herbals are generally perceived as safe, harmless, and free from side effects, but there have been reports of adulteration of Asian herbal remedies with Western pharmaceuticals, a practice that has not hitherto been observed in Africa. The authors present 2 cases from South Africa in which herbal remedies, consumed by patients admitted to hospital, were found to be adulterated with commercially available Western medicines. The adulterants were identified using high-performance liquid chromatography with a photodiode array detector and gas chromatography-mass spectrometry. The authors alert clinicians to the fact that adulteration of herbal remedies may become more common and should be considered, especially in cases where the symptoms are confusing in light of the history.