Merck
Cutaneous reactions to imatinib mesylate treated by topical steroid.
American journal of hematology (2005-02-24)
Hideyuki Ujiie, Tadamichi Shimizu, Hiroshi Shimizu
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
6α,9α-Difluoroprednisolone 21-acetate 17-butyrate, ≥98%