Merck
  • Stability-indicating methods for determination of pyritinol dihydrochloride in the presence of its precursor and degradation product by derivative spectrophotometry.
Stability-indicating methods for determination of pyritinol dihydrochloride in the presence of its precursor and degradation product by derivative spectrophotometry.
Journal of AOAC International (2005-03-12)
Mostafa A Shehata, Mohammad A el-Sayed, Mohammad G el-Bardicy, Mohammad F el-Tarras
ZUSAMMENFASSUNG
A first-derivative spectrophotometric (1D) method and a derivative-ratio zero-crossing spectrophotometric (1DD) method were used to determine pyritinol dihydrochloride (I) in the presence of its precursor (II) and its degradation product (III) with 0.1N hydrochloric acid as a solvent. Linear relationships were obtained in the ranges of 6-22 microg/mL for the (1D) method and 6-20 microg/mL for the (1DD) method. By applying the proposed methods, it was possible to determine pyritinol dihydrochloride in its pure powdered form with an accuracy of 100.36 +/- 1.497% (n = 9) for the (1D) method and an accuracy of 99.92 +/- 1.172% (n = 8) for the (1DD) method. Laboratory-prepared mixtures containing different ratios of (I), (II), and (III) were analyzed, and the proposed methods were valid for concentrations of < or = 10% (II) and < or = 50% (III). The proposed methods were validated and found to be suitable as stability-indicating assay methods for pyritinol in pharmaceutical formulations.