Merck
  • Design of small-sized libraries by combinatorial assembly of linkers and functional groups to a given scaffold: application to the structure-based optimization of a phosphodiesterase 4 inhibitor.
Design of small-sized libraries by combinatorial assembly of linkers and functional groups to a given scaffold: application to the structure-based optimization of a phosphodiesterase 4 inhibitor.
Journal of medicinal chemistry (2005-05-27)
Mireille Krier, João X de Araújo-Júnior, Martine Schmitt, Jérôme Duranton, Hélène Justiano-Basaran, Claire Lugnier, Jean-Jacques Bourguignon, Didier Rognan
ZUSAMMENFASSUNG
Combinatorial chemistry and library design have been reconciled by applying simple medicinal chemistry concepts to virtual library design. The herein reported "Scaffold-Linker-Functional Group" (SLF) approach has the aim to maximize diversity while minimizing the size of a scaffold-focused library with the aid of simple molecular variations in order to identify critical pharmacophoric elements. Straightforward rules define the way of assembling three building blocks: a conserved scaffold, a variable linker, and a variable functional group. By carefully selecting a limited number of functional groups not overlapping in pharmacophoric space, the size of the library is kept to a few hundred. As an application of the SLF approach, a small-sized combinatorial library (320 compounds) was derived from the scaffold of the known phosphodiesterase 4 inhibitor zardaverine. The most interesting analogues were further prioritized for synthesis and enzyme inhibition by FlexX docking to the X-ray structure of the enzyme, leading to a 900-fold increased affinity within nine synthesized compounds and a single screening round.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Zardaverine