Merck
  • Sequential conformation changes of chinese hamster ovary dihydrofolate reductase at its active sites.
Sequential conformation changes of chinese hamster ovary dihydrofolate reductase at its active sites.
Protein and peptide letters (2005-07-21)
Wen-he Zhang, San-bo Qin, Hong-jie Zhang, Zhi-yong Zhu
ZUSAMMENFASSUNG
The local fluorescence probes, 2-(p-toluidino)-6-naphthalenesulfonic acid (TNS) and NADPH were employed to detect urea-induced conformation changes at each active site of dihydrofolate reductase (DHFR), respectively. The results indicate that local conformation change at DHF/TNS could be superimposed by the conformation change calculated from the enzyme activity change with a three-state model; while at NADPH site it is lagged in the first transition. This difference is further supported by the different relative changes of Michaelis constants at 0, 1 and 1.8 M urea for each substrate. Our results suggest that local conformation at DHF site is more flexible than that at NADPH site, and the urea-induced unfolding could be ascribed to a four-state transition.