Merck
  • The screening of chemicals for juvenoid-related endocrine activity using the water flea Daphnia magna.
The screening of chemicals for juvenoid-related endocrine activity using the water flea Daphnia magna.
Aquatic toxicology (Amsterdam, Netherlands) (2005-07-28)
Helen Ying Wang, Allen W Olmstead, Hong Li, Gerald A Leblanc
ZUSAMMENFASSUNG
U.S. Environmental Protection Agency is charged with developing a screening and testing paradigm for detecting endocrine toxicity of chemicals that are subject to regulation under the Food Quality Protection and the Safe Drinking Water Acts. In this study, we developed and evaluated a screening assay that could be employed to detect juvenoid-related endocrine-modulating activity in an invertebrate species. Juvenoid activity, anti-juvenoid activity, and juvenoid potentiator activity of chemicals was assessed using the water flea Daphnia magna. Male sex determination is under the regulatory control of juvenoid hormone, presumably methyl farnesoate, and this endpoint was used to detect juvenoid modulating activity of chemicals. Eighteen chemicals were evaluated for juvenoid agonist activity. Positive responses were detected with the juvenoid hormones methyl farnesoate and juvenile hormone III along with the insect growth regulating insecticides pyriproxyfen, fenoxycarb, and methoprene. Weak juvenoid activity also was detected with the cyclodiene insecticide dieldrin. Assays performed repetitively with compounds that gave either strong positive, weak positive, or negative response were 100% consistent indicating that the assay is not prone to false positive or negative responses. Five candidate chemicals were evaluated for anti-juvenoid activity and none registered positive. Four chemicals (all trans-retinoic acid, methoprene, kinoprene, bisphenol A) also were evaluated for their ability to potentiate the activity of methyl farnesoate. All registered positive. Results demonstrate that an in vivo assay with a crustacean species customarily employed in toxicity testing can be used to effectively screen chemicals for juvenoid-modulating activity.