Merck
  • Disposition of cocaine and its metabolites in human sweat after controlled cocaine administration.
Disposition of cocaine and its metabolites in human sweat after controlled cocaine administration.
Clinical chemistry (2005-09-17)
Sherri L Kacinko, Allan J Barnes, Eugene W Schwilke, Edward J Cone, Eric T Moolchan, Marilyn A Huestis
ZUSAMMENFASSUNG
Sweat testing is a noninvasive technique for monitoring drug exposure in treatment, criminal justice, and employment settings. We evaluated cocaine excretion in 9 participants' sweat after they received 3 low doses (75 mg/70 kg) of cocaine HCl subcutaneously within 1 week and, 3 weeks later, 3 high doses (150 mg/70 kg). Six additional participants completed portions of the study. PharmChek sweat patches (n = 1390) were collected throughout a 3-week washout period, reflecting previously self-administered drugs, and during and after controlled dosing. Cocaine was the primary analyte detected with 24% of patches positive at the gas chromatography-mass spectrometry limit of quantification of 2.5 ng/patch and 7% of patches at the proposed Substance Abuse and Mental Health Services Administration cutoff of 25 ng/patch. Ecgonine methyl ester (EME) was detected more often and at generally higher concentrations than benzoylecgonine. In patches containing both metabolites, there was no statistically significant difference in the benzoylecgonine/EME ratio based on length of patch wear. During washout, 2 participants' weekly patches tested positive (> or =25 ng/patch) during the first week; one remained positive during week 2; and none were positive during week 3. Cocaine and EME were detectable within 2 h; benzoylecgonine was not detected until 4-8 h after low doses and slightly sooner after high doses. The majority of drug was excreted within 24 h. Over 70% of weekly patches worn during low doses were positive for cocaine (> or =25 ng/patch), increasing to 100% during high doses. Sweat testing is an effective and reliable method of monitoring cocaine exposure.