Merck
  • Antigen-induced Ca2+ mobilization in RBL-2H3 cells: role of I(1,4,5)P3 and S1P and necessity of I(1,4,5)P3 production.
Antigen-induced Ca2+ mobilization in RBL-2H3 cells: role of I(1,4,5)P3 and S1P and necessity of I(1,4,5)P3 production.
Cell calcium (2005-10-13)
Hyun-Sil Lee, Chang-Shin Park, Young Mi Lee, Ho Young Suk, Tameka C M Clemons, Oksoon Hong Choi
ZUSAMMENFASSUNG
Inositol 1,4,5-trisphosphate (IP3) has long been recognized as a second messenger for intracellular Ca2+ mobilization. Recently, sphingosine 1-phosphate (S1P) has been shown to be involved in Ca2+ release from the endoplasmic reticulum (ER). Here, we investigated the role of S1P and IP3 in antigen (Ag)-induced intracellular Ca2+ mobilization in RBL-2H3 mast cells. Antigen-induced intracellular Ca2+ mobilization was only partially inhibited by the sphingosine kinase inhibitor dl-threo-dihydrosphingosine (DHS) or the IP3 receptor inhibitor 2-aminoethoxydiphenyl borate (2-APB), whereas preincubation with both inhibitors led to complete inhibition. In contrast, stimulation of A3 adenosine receptors with N5-ethylcarboxamidoadenosine (NECA) caused intracellular Ca2+ mobilization that was completely abolished by 2-APB but not by DHS, suggesting that NECA required only the IP3 pathway, while antigen used both the IP3 and S1P pathways. Interestingly, however, inhibition of IP3 production with the phospholipase C inhibitor U73122 completely abolished Ca2+ release from the ER induced by either stimulant. This suggested that S1P alone, without concomitant production of IP3, would not cause intracellular Ca2+ mobilization. This was further demonstrated in some clones of RBL-2H3 cells excessively overexpressing a beta isoform of Class II phosphatidylinositol 3-kinase (PI3KC2beta). In such clones including clone 5A4C, PI3KC2beta was overexpressed throughout the cell, although endogenous PI3KC2beta was normally expressed only in the ER. Overexpression of PI3KC2beta in the cytosol and the PM led to depletion of phosphatidylinositol 4,5-bisphosphate (PI(4,5)P2), resulting in a marked reduction in IP3 production. This could explain the abolishment of intracellular Ca2+ mobilization in clone 5A4C. Supporting this hypothesis, the Ca2+ mobilization was reconstituted by the addition of exogenous PI(4,5)P2 in these cells. Our results suggest that both IP3 and S1P contribute to FcvarepsilonRI-induced Ca2+ release from the ER and production of IP3 is necessary for S1P to cause Ca2+ mobilization from the ER.