Merck
Trypanocidal activity of oleoresin and terpenoids isolated from Pinus oocarpa.
Zeitschrift fur Naturforschung. C, Journal of biosciences (2005-12-03)
Julieta Rubio, José S Calderón, Angélica Flores, Clementina Castroa, Carlos L Céspedes
ZUSAMMENFASSUNG
Fractionation with n-hexane/ethyl acetate (1:1 v/v) by open column chromatography of the oleoresin from Pinus oocarpa Schiede yielded two diterpenes, pimaric acid (1) and dehydroabietic acid (5), the sesquiterpene longifolene (3) and a diterpenic mixture containing pimaric acid (1), isopimaric acid (4) and dehydroabietic acid (5). Subsequently, the isolated compounds, the mixture of 1, 4 and 5, the oleoresin and the dehydroabietic acid methyl ester (2), were tested in vitro against epimastigotes of Trypanosoma cruzi, the causative agent of Chagas disease. The most active compounds were 1, 3 and the oleoresin, being as active as nifurtimox, a drug effective in the treatment of acute infection by American trypanosomiasis and used in this work as positive control.