Merck
  • Selective inhibition of fibroblast activation protein protease based on dipeptide substrate specificity.
Selective inhibition of fibroblast activation protein protease based on dipeptide substrate specificity.
The Journal of biological chemistry (2006-01-18)
Conrad Yap Edosada, Clifford Quan, Christian Wiesmann, Thuy Tran, Dan Sutherlin, Mark Reynolds, J Michael Elliott, Helga Raab, Wayne Fairbrother, Beni B Wolf
ZUSAMMENFASSUNG
Fibroblast activation protein (FAP) is a transmembrane serine peptidase that belongs to the prolyl peptidase family. FAP has been implicated in cancer; however, its specific role remains elusive because inhibitors that distinguish FAP from other prolyl peptidases like dipeptidyl peptidase-4 (DPP-4) have not been developed. To identify peptide motifs for FAP-selective inhibitor design, we used P(2)-Pro(1) and acetyl (Ac)-P(2)-Pro(1) dipeptide substrate libraries, where P(2) was varied and substrate hydrolysis occurs between Pro(1) and a fluorescent leaving group. With the P(2)-Pro(1) library, FAP preferred Ile, Pro, or Arg at the P(2) residue; however, DPP-4 showed broad reactivity against this library, precluding selectivity. By contrast, with the Ac-P(2)-Pro(1) library, FAP cleaved only Ac-Gly-Pro, whereas DPP-4 showed little reactivity with all substrates. FAP also cleaved formyl-, benzyloxycarbonyl-, biotinyl-, and peptidyl-Gly-Pro substrates, which DPP-4 cleaved poorly, suggesting an N-acyl-Gly-Pro motif for inhibitor design. Therefore, we synthesized and tested the compound Ac-Gly-prolineboronic acid, which inhibited FAP with a K(i) of 23 +/- 3 nm. This was approximately 9- to approximately 5400-fold lower than the K(i) values for other prolyl peptidases, including DPP-4, DPP-7, DPP-8, DPP-9, prolyl oligopeptidase, and acylpeptide hydrolase. These results identify Ac-Gly-BoroPro as a FAP-selective inhibitor and suggest that N-acyl-Gly-Pro-based inhibitors will allow testing of FAP as a therapeutic target.