Merck
Raynaud phenomena and migraine in two children: inclusion within a family of related disorders.
Acta paediatrica (Oslo, Norway : 1992) (2006-01-20)
Rachel Ej Besser, Colin Mm Stern
ZUSAMMENFASSUNG
We report a case of two siblings with Raynaud phenomena and migraine, whose symptoms were successfully treated with pizotifen. To our knowledge, this is the first case documenting the association between Raynaud phenomena and migraine in two siblings with a family history of Raynaud phenomena and ischaemic heart disease.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Pizotifen, ≥98% (HPLC)