- The influence of superoxide on the production of hypochlorous acid by human neutrophils.
The influence of superoxide on the production of hypochlorous acid by human neutrophils.
Free radical research communications (1991-01-01)
A J Kettle, C C Winterbourn
PMID1649101
ZUSAMMENFASSUNG
Human neutrophils stimulated with opsonized zymosan promoted hypochlorous acid (HOCl)-dependent loss of monochlorodimedon. Formation of HOCl was completely inhibited by catalase, and it was also inhibited up to 70% by SOD. There was no inhibition by desferal, DTPA, mannitol or dimethylsulphoxide, which excluded the involvement of .OH. Our results indicate that generation of O2- by neutrophils enables these cells to enhance their production of HOCl. Furthermore, inhibition of neutrophil processes by SOD and catalase does not necessarily implicate .OH. We propose that O2- may potentiate oxidant damage at inflammatory sites by boosting the myeloperoxidase-dependent production of HOCl.