Merck
  • Pathogenicity of Salmonella: SopE-mediated membrane ruffling is independent of inositol phosphate signals.
Pathogenicity of Salmonella: SopE-mediated membrane ruffling is independent of inositol phosphate signals.
FEBS letters (2006-02-28)
Sandrine Deleu, Kuicheon Choi, Jeff M Reece, Stephen B Shears
ZUSAMMENFASSUNG
Studies [Zhou, D., Chen, L.-M., Hernandez, L., Shears, S.B., and Galán, J.E. (2001) A Salmonella inositol polyphosphatase acts in conjunction with other bacterial effectors to promote host-cell actin cytoskeleton rearrangements and bacterial internalization. Mol. Microbiol. 39, 248-259] with engineered Salmonella mutants showed that deletion of SopE attenuated the pathogen's ability to deplete host-cell InsP5 and remodel the cytoskeleton. We pursued these observations: In SopE-transfected host-cells, membrane ruffling was induced, but SopE did not dephosphorylate InsP5, nor did it recruit PTEN (a cytosolic InsP5 phosphatase) for this task. However, PTEN strengthened SopE-mediated membrane ruffling. We conclude SopE promotes host-cell InsP5 hydrolysis only with the assistance of other Salmonella proteins. Our demonstration that Salmonella-mediated cytoskeletal modifications are independent of inositolphosphates will focus future studies on elucidating alternate pathogenic consequences of InsP5 metabolism, including ion channel conductance and apoptosis.