Merck
  • Effect of catechol oestrogens on extraneuronal metabolism of noradrenaline by rabbit uterine endometrium and myometrium.
Effect of catechol oestrogens on extraneuronal metabolism of noradrenaline by rabbit uterine endometrium and myometrium.
Naunyn-Schmiedeberg's archives of pharmacology (1991-03-01)
J A Kennedy
ZUSAMMENFASSUNG
The effects of 2-hydroxy and 2-methoxy oestrogens on the extraneuronal O-methylation of 3H-(-)-noradrenaline were examined in progesterone-dominated, monoamine oxidase (MAO)-inhibited, rabbit uterine tissues in vitro. Both the corticosteroid-sensitive system in myometrium and the cocaine-sensitive system in endometrium were examined. In myometrial slices preincubated with nialamide to inhibit MAO and incubated with cocaine to inhibit neuronal uptake, 3H-normetanephrine (3H-NMN) formation was inhibited in the order of potency 2-hydroxy oestrone greater than or equal to 2-hydroxy oestradiol = 2-methoxy oestradiol greater than or equal to 2-methoxy oestrone. In myometrial slices not exposed to cocaine and nialamide, inhibition of 3H-NMN formation by both 2-hydroxy and 2-methoxy oestradiol did not affect the formation of deaminated metabolites of 3H-(-)-noradrenaline by the alternative metabolising pathway. In endometrial slices preincubated with nialamide to inhibit MAO, only 2-hydroxy oestrogens inhibited 3H-NMN formation, but they were one to two orders of magnitude less potent in this regard than in the myometrium. The uptake of 3H-(-)-noradrenaline by MAO- and COMT-inhibited myometrial slices was inhibited by 2-hydroxy and 2-methoxy oestrogens in the order of potency 2-methoxy oestradiol greater than or equal to 2-methoxy oestrone greater than or equal to 2-hydroxy oestrone greater than 2-hydroxy oestradiol. Uptake of 3H-(-)-noradrenaline by endometrial slices was not affected by either 2-hydroxy or 2-methoxy oestrogens.(ABSTRACT TRUNCATED AT 250 WORDS)