Merck
  • Enantioselective synthesis of a novel carbocyclic C-nucleoside via asymmetric Diels-Alder reaction of (S)s-3-(2-pyridylsulfinyl)-acrylate.
Enantioselective synthesis of a novel carbocyclic C-nucleoside via asymmetric Diels-Alder reaction of (S)s-3-(2-pyridylsulfinyl)-acrylate.
Nucleic acids symposium series (1991-01-01)
T Takahashi, H Kotsubo, T Koizumi