Merck
  • Effects of the hypocholesterolemic drug, 4-aminopyrazolo (3,4-d) pyrimidine (4-APP), on the hamster adrenal cortex. An ultrastructural and functional study.
Effects of the hypocholesterolemic drug, 4-aminopyrazolo (3,4-d) pyrimidine (4-APP), on the hamster adrenal cortex. An ultrastructural and functional study.
Virchows Archiv. B, Cell pathology including molecular pathology (1991-01-01)
A Kasprzak, P Rebuffat, J B Warchol, G Mazzocchi, G G Nussdorfer, L K Malendowicz
ZUSAMMENFASSUNG
The aim of this study was to gain insight into the effects of 4-aminopyrazolo(3,4-d)pyrimidine (4-APP), a hypocholesterolemic drug, on the adrenal cortex of the hamster, representing an animal species in which steroidogenesis primarily relies on utilization of cholesterol synthesized de novo in the gland. 4-APP administration (1.5 mg/animal day for 3 days) to intact or dexamethasone-suppressed hamsters resulted in a marked proliferation of adrenocortical cells. However, the volume of parenchymal cells was unchanged in intact animals and lowered in the zona glomerulosa (ZG) and zona reticularis (ZR) of dexamethasone-administered hamsters. In both groups of animals, 4-APP strikingly increased the volume of the lipid-droplet compartment and markedly reduced the surface area of smooth endoplasmic reticulum in ZF cells, without significantly affecting the volume of the mitochondrial compartment and the surface area of mitochondrial cristae. These morphologic changes displayed no evident correlation with adrenal cortisol content and secretion. Since most of the 4-APP-induced changes were not prevented by dexamethasone, it seems legitimate to suggest that they could mainly depend on a direct effect of 4-APP on the hamster adrenocortical cells.