Merck
Adverse extrapyramidal effects in four horse given fluphenazine decanoate.
Journal of the American Veterinary Medical Association (2006-07-05)
John D Baird, Luis G Arroyo, Modest Vengust, M Kimberly J McGurrin, Alexander Rodriguez-Palacios, Daniel G Kenney, Manickam Aravagiri, George A Maylin
ZUSAMMENFASSUNG
4 racehorses were examined because of markedly abnormal behavior following administration of fluphenazine decanoate. Clinical signs included restlessness, agitation, profuse sweating, hypermetria, aimless circling, intense pawing and striking with the thoracic limbs, and rhythmic swinging of the head and neck alternating with episodes of severe stupor. Fluphenazine was detected in serum or plasma from all 4 horses. The dose of fluphenazine decanoate administered to 3 of the 4 horses was within the range (25 to 50 mg) routinely administered to adult humans. In 2 horses, there was no response to IV administration of diphenhydramine hydrochloride, but the abnormal behavior in these 2 horses appeared to resolve following administration of benztropine mesylate, and both horses returned to racing. The other 2 horses responded to diphenhydramine administration. One returned to racing. The other was euthanized because of severe neurologic signs, respiratory failure, and acute renal failure. Findings indicate that adverse extrapyramidal effects may occur in horses given fluphenazine decanoate. These effects appear to be unpredictable and may be severe and life threatening. Use of fluphenazine decanoate as an anxiolytic in performance horses is not permitted in many racing and horse show jurisdictions, and analytic procedures are now available to detect the presence of fluphenazine in serum or plasma.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Fluphenazindecanoat, European Pharmacopoeia (EP) Reference Standard