- Himbacine derived thrombin receptor (PAR-1) antagonists: structure-activity relationship of the lactone ring.
Himbacine derived thrombin receptor (PAR-1) antagonists: structure-activity relationship of the lactone ring.
Bioorganic & medicinal chemistry letters (2006-07-11)
Yan Xia, Samuel Chackalamannil, Tze-Ming Chan, Michael Czarniecki, Darío Doller, Keith Eagen, William J Greenlee, Hsingan Tsai, Yuguang Wang, Ho-Sam Ahn, George C Boykow, Andrew T McPhail
PMID16824760
ZUSAMMENFASSUNG
The structure-activity relationship (SAR) of the lactone ring of himbacine derived thrombin receptor (PAR-1) antagonists (e.g., 2-5) is described. The effect of the lactone carbonyl group on binding to PAR-1 is dependent on the substitution pattern of the pyridine ring. A stereoselective intramolecular Michael addition reaction to the vinyl pyridine group was observed for these pyridine analogs of himbacine in basic conditions at elevated temperature.