Merck
  • An orally active selective androgen receptor modulator is efficacious on bone, muscle, and sex function with reduced impact on prostate.
An orally active selective androgen receptor modulator is efficacious on bone, muscle, and sex function with reduced impact on prostate.
Endocrinology (2006-10-07)
Jeffrey N Miner, William Chang, Mark S Chapman, Patricia D Finn, Mei Hua Hong, Francisco J López, Keith B Marschke, Jon Rosen, William Schrader, Russell Turner, Arjan van Oeveren, Humberto Viveros, Lin Zhi, Andres Negro-Vilar
ZUSAMMENFASSUNG
A number of conditions, including osteoporosis, frailty, and sexual dysfunction in both men and women have been improved using androgens. However, androgens are not widely used for these indications because of the side effects associated with these drugs. We describe an androgen receptor (AR) ligand that maintains expected anabolic activities with substantially diminished activity in the prostate. LGD2226 is a nonsteroidal, nonaromatizable, highly selective ligand for the AR, exhibiting virtually no affinity for the other intracellular receptors. We determined that AR bound to LGD2226 exhibits a unique pattern of protein-protein interactions compared with testosterone, fluoxymesterone (an orally available steroidal androgen), and other steroids, suggesting that LGD2226 alters the conformation of the ligand-binding domain. We demonstrated that LGD2226 is fully active in cell-based models of bone and muscle. LGD2226 exhibited anabolic activity on muscle and bone with reduced impact on prostate growth in rodent models. Biomechanical testing of bones from animals treated with LGD2226 showed strong enhancement of bone strength above sham levels. LGD2226 was also efficacious in a sex-behavior model in male rats measuring mounts, intromissions, ejaculations, and copulation efficiency. These results with an orally available, nonaromatizable androgen demonstrate the important role of the AR and androgens in mediating a number of beneficial effects in bone, muscle, and sexual function independent from the conversion of androgens into estrogenic ligands. Taken together, these results suggest that orally active, nonsteroidal selective androgen receptor modulators may be useful therapeutics for enhancing muscle, bone, and sexual function.