- New hydroxyl protecting groups of a safety-catch type removable by reductive acidolysis.
New hydroxyl protecting groups of a safety-catch type removable by reductive acidolysis.
Chemical & pharmaceutical bulletin (1991-11-01)
Y Kiso, S Tanaka, T Kimura, H Itoh, K Akaji
PMID1724744
ZUSAMMENFASSUNG
New hydroxyl protecting groups of a safety-catch type, i.e., 4-methylsulfinylbenzyl-oxycarbonyl (Msz) group for Tyr and 4-methylsulfinylbenzyl (Msob) ether for Thr, have been developed. O-Msz and O-Msob groups are stable under both acidic and basic conditions and can be removed by a one-pot reaction involving reductive acidolysis using tetrachlorosilane-trifluoroacetic acid (TFA)-scavengers. Using these new protecting groups, a 17 residue-peptide, gamma-endorphin, was successfully synthesized by the efficient solid phase method.