Merck
  • Gender difference in the pharmacokinetic interaction between oral warfarin and oxolamine in rats: inhibition of CYP2B1 by oxolamine in male rats.
Gender difference in the pharmacokinetic interaction between oral warfarin and oxolamine in rats: inhibition of CYP2B1 by oxolamine in male rats.
Biopharmaceutics & drug disposition (2007-02-14)
Xuan Zhu, Dae Y Lee, Wan G Shin
ZUSAMMENFASSUNG
The possible reason for the significantly greater AUC of oral warfarin with oral oxolamine in male Sprague-Dawley rats was evaluated. After oral administration of warfarin at a dose of 2 mg/kg to male rats with oxolamine at doses of 10 and 50 mg/kg, the AUC values of warfarin were significantly greater than the controls (254 and 330 versus 180 microg h/ml). However, the AUC values of warfarin were not affected by oxolamine in female rats. This could be due to inhibition of CYP2B1, 2C11 and 3A2 by oxolamine in male rats, since warfarin was metabolized via CYP1A1, 2B1, 2C6, 2C11 and 3A2 in rats and CYP2B1 is male dominant, and CYP2C11 and 3A2 are male specific. Therefore, phenytoin, torasemide and clarithromycin (mainly metabolized via CYP2B1/2, 2C11 and 3A2 in rats, respectively) were administered intravenously to male rats with or without oral oxolamine. After oral oxolamine at doses of 10 and 50 mg/kg, the AUC of phenytoin was significantly greater (1280 and 1640 versus 938 microg min/ml), however, the AUC values of torasemide and clarithromycin were independent of oxolamine. The above data suggest that the significantly greater AUC of oral warfarin with oral oxolamine could be due to inhibition of CYP2B1/2 by oxolamine in male rats.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Oxolamin -citrat (Salz)